All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
isotopic purity
99 atom % 13C
optical activity
[α]20/D −20°, c = 4 in H2O
mp
94 °C (dec.) (lit.)
SMILES string
OC[C@H]1O[13CH](O)[C@H](O)[C@@H]1O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service