Skip to Content
Merck
All Photos(1)

Key Documents

153656

Sigma-Aldrich

1,2,3,5-Tetramethylbenzene

80%

Synonym(s):

Isodurene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(CH3)4
CAS Number:
Molecular Weight:
134.22
Beilstein:
1904391
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

80%

refractive index

n20/D 1.512 (lit.)

density

0.891 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c(C)c(C)c1

Looking for similar products? Visit Product Comparison Guide

Other Notes

Remainder 1,2,4,5-tetramethylbenzene

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service