CHO cell culture media provides all the nutrients required for cellular growth and metabolism suck as amino acids, buffers, growth factors, vitamins, and lipids. Growth factors like insulin and insulin analogs are critical for cell growth, proliferation, and higher productivity.
Cysteine and Tyrosine are critical for CHO cells which need to be added separately due to poor solubility at neutral PH. We provide non-animal origin GMP-certified growth factors (supplements) and modified amino acids like phosphotyrosine and sulfo cysteine which are soluble in neutral feeds and can be fed along with main feed for upstream fed-batch process simplification.
In this webinar, you will learn about:
- CHO cell culture media components
- Non-animal origin growth factors
- The usage of modified amino acids for fed-batch simplification
Speaker
![Sateesh Poojari](/deepweb/assets/sigmaaldrich/marketing/global/images/webinars/satish-poojari/satish-poojari.jpg)
Sateesh Poojari
Merck
Sr. Technical Application Expert
Satish has 17 years of experience in research, sales, sales development, field marketing, and technology management. He is an expert in cell line development, upstream process development using E. coli, and CHO platforms for expressing biotherapeutics and biosimilars. For the past 7 years, he has worked with the Technical University Munich, Hetero, Aurobindo, and others associated with Merck, supporting our bioprocess business in India. Sateesh holds a double masters, one in biotechnology from Bangalore University and one in molecular biology from the University of Skovde, Sweden.
Pharma and biopharma manufacturing
- Pharma and biopharma manufacturing
Duration:1h
Language:English
Session 1:presented May 2, 2023