CRISPRi & CRISPRa Genetic Screening
Lentiviral Libraries and Pools for CRISPR Activation and Inhibition
Discover our optimized CRISPR activation and CRISPR inhibition libraries for gene knockdown and overexpression experiments. Our pooled CRISPRi and SAM CRISPRa lentiviral libraries are available off-the-shelf or customized. Individual guide RNAs for both systems are also easily ordered online.
Sigma-Aldrich® Advanced Genomics CRISPRi and CRISPRa libraries complement our comprehensive suite of genome engineering technologies to allow you to uncover further insights into complex biological processes. Large scale LOF (loss-of-function) and GOF (gain-of-function) screens can identify unique, yet functionally related, gene pathways and hits which are often missed under otherwise identical KO (knockout) screening parameters. Take your experiments Beyond the Bench with the largest genomics portfolio available and a partner that can ensure your experimental goals are met.
Specifications | |
---|---|
CRISPRi | CRISPRa |
|
|
The Sigma-Aldrich® CRISPR inhibition libraries have superior algorithm placement and improved sgRNA scaffolding for optimal gene knockdown, even in the most difficult targets.
Added Benefits:
- We are a 10x Genomics compatible partner with ready-to-use custom CRISPR lenti pools with Feature Barcode technology included in our vectors to enable discovery at single-cell resolution
- Individual CRISPR inhibition & activation guides can be easily ordered online – predesigned or custom
- In house DNA Prep and Deconvolution Services
Figure 1.CRISPRi and CRISPRa utilize catalytically inactive dCas9 to recruit the transcriptional repressor KRAB domain or the transcriptional activators HSF1, p65 and MS2 to silence and overexpress target gene expression respectively.
Not sure which screening tool is right for your experiment? Pricing questions? Need a custom pool? We can help!
Resources
Vector Maps
SAM CRISPRa
CRISPRi
CRISPRi Available Libraries and Pools
Product Description | Target | Number of Genes | Product Number |
---|---|---|---|
Whole-Genome Libraries | |||
Human Whole Genome & Non-Coding CRISPRi Library | Human, Whole genome | 18,905 | CRISPRILIB |
Control & Effector Kit: Positive & Negative controls with dCas9 | Coding & non-targeting | Coding gene RAB1A and a non-targeting control | CRISPRICON |
CRISPRi Subpools | |||
10x Genomics Feature Barcode Optimization Kit | Human, Coding | Coding gene RAB1A and a non-targeting control | CRISPRI10X |
Druggable Genome Pool | Human, Coding | 2,318 | CRISPRIS01 |
Cancer & Apoptosis Pool | Human, Coding | 2,916 | CRISPRIS02 |
Stress Proteostasis Pool | Human, Coding> | 3,093 | CRISPRIS03 |
Mitochondria/Trafficking/Motility Pool | Human, Coding | 2,219 | CRISPRIS04 |
Gene Expression Pool (Transcription Factors) | Human, Coding | 2,292 | CRISPRIS05 |
Membrane Proteins Pool | Human, Coding | 2,418 | CRISPRIS06 |
Unassigned Pool (Uncategorized Gene Function, Novel Targets) | Human, Coding | 1,824 | CRISPRIS07 |
Common Cancer Cell Lines Pool & iPSC Pool: Common to 7 cell lines (5 cancer, iPSC, HFF) | Human, Long non-coding | 691 | CRISPRIS08 |
iPSC Pool | Human, Long non-coding | 3,000 | CRISPRIS09 |
Common Cancer Cell Lines Pool (HeLa, HEK293T, K562, MCF7, U87 cell lines) | Human, Long non-coding | 1,329 | CRISPRIS10 |
SAM CRISPRa Libraries and Helper Constructs
Product Description | Target | Number of Genes | Product Number |
---|---|---|---|
Whole-Genome SAM Libraries | |||
SAM CRISPRa Library Zeo | Human, coding | 23,430 | HSAMZEO |
SAM CRISPRa Library Puro | Human, coding | 23,430 | HSAMPURO |
SAM CRISPRa Library Zeo | Mouse, coding | 23,439 | MSAMZEO |
SAM CRISPRa Library Puro | Mouse, coding | 23,439 | MSAMPURO |
SAM Helper Constructs: Plasmid DNA | |||
dCas9-VP64-Blasticidin SAM Helper Construct 1 | Mammalian | SAMVP64BST | |
MS2-P65-HSF1-Hygromycin SAM Helper Construct 2 | Mammalian | SAMMS2HYG | |
SAM Helper Construct Kit | Mammalian | SAMHELPERP | |
SAM Helper Constructs: Lentivirus | |||
dCas9-VP64-Blasticidin SAM Helper Construct 1 | Mammalian | SAMVP64BSTV | |
MS2-P65-HSF1-Hygromycin SAM Helper Construct 2 | Mammalian | SAMMS2HYGV | |
SAM Helper Construct Kit | Mammalian | SAMHELPERV |
CRISPR Screening Tips
A simple modification to improve CRISPR-pooled screenin result. Reduce NGS noise by eliminating artifact DNA.