385906
Hyaluronic Acid, Bovine Trachea, Fluorescein-Labeled
Fluorescently-labeled hyaluronic acid containing approximately 5 mole-percent of coupled dye. Useful for binding and localization studies.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352201
NACRES:
NA.79
Recommended Products
Quality Level
Assay
≥95% (NMR)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
orange
solubility
water: 1 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
Fluorescently-labeled hyaluronic acid containing approximately 5 mole-percent of coupled dye. Useful for binding and localization studies.
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Asari, A., et al. 1998. Arch. Histol. Cytol. 61, 125.
Cantor, J.O., et al. 1997. Exp. Lung Res. 23, 229.
Cantor, J.O., et al. 1997. Exp. Lung Res. 23, 229.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service