CLS6795410D
Corning® stirrer with digital display
AC/DC input 120 V AC, US 3-pin plug, plate L × W 5 in. × 7 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
All Photos(1)
About This Item
UNSPSC Code:
41103803
NACRES:
NB.73
Recommended Products
packaging
case of 1
manufacturer/tradename
Corning 6795-410D
availability
not available in EU
parameter
60-1150 rpm speed
plate L × W
5 in. × 7 in.
Related Categories
General description
- Digital LED stir speed display indicates when set speed has been reached
- Microprocessor maintains consistent and repeatable stir speed settings
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
Other Notes
individual units are not routinely calibrated, rpm speeds are approximate only
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)