MT30046248
Mettler Toledo SevenExcellence™ pH/Ion meter S500
S500 basic (no electrode), AC/DC input 230 V AC, universal plug set
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115603
NACRES:
NB.61
description
Concentration range 0.001 … 999 999 mg/L, ppm ; 0.0001 … 100 mol/L, % ; 0.001 … 100 000 mmol/L (+/- 0.5%)
manufacturer/tradename
Mettler-Toledo 30046248
mV measuring range
-2000-2000, accuracy: ±1
temperature measuring range
-30-130 °C, accuracy: ±0.1
pH range
-2-20
Looking for similar products? Visit Product Comparison Guide
General description
Includes meter with a pH/Ion expansion unit and 2 blank expansion units
Mettler Toledo SevenExcellence™ pH/Ion meter – intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and solid compliance. This single channel instrument is modularly expandable with additional measurement parameters at any time!
Included in every SevenExcellence:
uPlace™ electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
- Methods for incremental ion measurements, guiding the user through the individual steps
- Large 7 inch color display and intuitive menu guidance in 10 languages
- Flexible method concept for high reproducibility and security
- Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
- Integrated interfaces (USB, RS232 and Ethernet) for data exchange
- Comprehensive service package including IQ/OQ/PQ
Included in every SevenExcellence:
uPlace™ electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service