Skip to Content
Merck
All Photos(1)

Key Documents

Z614181

Vivaspin 2 centrifugal concentrators

MWCO 5 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24
material:
polycarbonate housing
polyethersulfone membrane
pore size:
5 kDa MWCO
dead stop volume:
8 μL
membrane area:
1.2 cm2
tube size:
~15 mL
working volume:
2 mL
sterility:
non-sterile
packaging:
package of 25 ea

material

polycarbonate housing
polyethersulfone membrane

sterility

non-sterile

feature

holdup volume 10 μL

packaging

package of 25 ea

manufacturer/tradename

Sartorius VS0211

parameter

8000 × g max. centrifugal force

dead stop volume

8 μL

membrane area

1.2 cm2

tube size

~15 mL

working volume

2 mL

pore size

5 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

The Vivaspin 2 combines the speed of the classic Vivaspin products with low internal surface and membrane area for superior recoveries from very dilute solutions. Unique to the Vivaspin 2 is the choice of directly pipetting the concentrate from the dead stop pocket built into the bottom of the concentrator, or alternatively reverse spinning into the concentrate recovery cap which can then be sealed for storage. Both methods result in near total concentrate recoveries.

  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service