Skip to Content
Merck
All Photos(1)

Key Documents

872100C

Avanti

18:1 PE-N-20:4

Avanti Research - A Croda Brand 872100C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-arachidonoyl (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H111N2O9P
CAS Number:
Molecular Weight:
1047.52
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (872100C-10mg)
pkg of 1 × 1 mL (872100C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 872100C

concentration

10 mg/mL (872100C-10mg)
5 mg/mL (872100C-5mg)

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C61H108NO9P.H3N/c1-4-7-10-13-16-19-22-25-28-29-32-33-36-39-42-45-48-51-59(63)62-54-55-69-72(66,67)70-57-58(71-61(65)53-50-47-44-41-38-35-31-27-24-21-18-15-12-9-6-3)56-68-60(64)52-49-46-43-40-37-34-30-26-23-20-17-14-11-8-5-2;/h16,19,25-28,30-33,39,42,58

Application

18:1 PE-N-20:4 has been used to spike N-acylphosphatidylethanolamine standard post liquid-liquid and solid-phase extraction by mass spectrometry. It has also been used as an internal standard to analyze the efficacy of CH3NH2-mediated deacylation.

Packaging

5 mL Clear Glass Sealed Ampule (872100C-10mg)
5 mL Clear Glass Sealed Ampule (872100C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative analysis of N-acylphosphatidylethanolamine molecular species in rat brain using solid-phase extraction combined with reversed-phase chromatography and tandem mass spectrometry
Triebl A, et al.
Journal of Separation Science, 39(13), 2474-2480 (2016)
A liquid chromatography-tandem mass spectrometry method for measurement of N-modified phosphatidylethanolamines
Guo L, et al.
Analytical Biochemistry, 405(2), 236-245 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service