Merck
Prolactin serum concentrations during aripiprazole treatment in youth.
Journal of child and adolescent psychopharmacology (2013-05-08)
Daniel J Safer, Chadi A Calarge, Alan M Safer
ABSTRACT
This study aimed to: document the extent of the reduction of serum prolactin (PRL) levels induced by aripiprazole (ARI) treatment in children and adolescents, compare this effect by age group, and shed light on this phenomenon. PRL serum levels in unmedicated subjects were compared to those in subjects treated with aripiprazole to calculate the rate of subnormal PRL levels during aripiprazole treatment. Next, a literature search was performed to better understand the effects of dopaminergic drugs on PRL levels by age group. Sixty percent of those treated with aripiprazole exhibited subnormal PRL serum levels versus 8% of unmedicated subjects. The rate of PRL subnormality in response to aripiprazole was half as frequent in adolescents and was minimal in adults. The drug-induced reduction of PRL serum levels became more prominent with increasing doses of aripiprazole and with an increased treatment duration. With the increasing use of aripiprazole in the United States population, it is important that future research be conducted to explore the potential sequelae of subnormal PRL serum levels in children and adolescents.
Product Description
Aripiprazole, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.