WPB01613WA
Whirl-Pak® Sponge Probe Bag
710 mL capacity(24 oz), pack of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Recommended Products
material
white polypropylene bag
sterility
sterile
feature
autoclavable: no
label plain
packaging
pack of 100 ea
manufacturer/tradename
Nasco Whirl-pak® B01613
bag L × W × thickness
9 in. (23 cm) × 6 in. (15 cm) × 0.64 mm
probe L
8 in. (20.3 cm)
capacity
710 mL capacity(24 oz)
Related Categories
General description
The Whirl-Pak® Sponge Probe is designed for surface sampling and is effective for hard-to-reach areas. The sponge is adhered to a holder that is screwed onto a handle (both pieces are made of polypropylene). The probe is sterilized with ethylene oxide gas inside a 24-oz. (710 ml) write-on bag. To use, add buffer to the bag, squeeze out the excess, swab the surface, insert the sponge back into the bag and, while holding it from the outside, unscrew the handle and discard. Then drop the sponge into the bag, close, and send to a lab for testing. The probe measures 8" (20.3 cm) long. foil pouch of of 100 bags.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service