Key Documents
EP5895101003
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x1000, to hold, 4 x 1,000 mL buckets, without buckets
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 4 x 1,000 mL buckets
to hold 4 x plate/tube buckets
swing-bucket rotor
without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895101003 (Global)
parameter
3,153 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5920R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG