Skip to Content
Merck
All Photos(1)

Documents

8.14676

Sigma-Aldrich

4,4′-Methylenebis(3-chloro-2,6-diethylaniline)

for synthesis

Synonym(s):

4,4′-Methylenebis(3-chloro-2,6-diethylaniline), Bis-(4-amino-2-chloro-3,5-diethylphenyl)-methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28Cl2N2
CAS Number:
Molecular Weight:
379.37
UNSPSC Code:
12352116
EC Index Number:
402-130-7
NACRES:
NA.22

vapor pressure

0.01 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (HPLC)

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

88-90 °C

solubility

slightly soluble 0.02 mg/L

density

1.24 g/cm3 at 20 °C

bulk density

610‑650 kg/m3

storage temp.

2-30°C

InChI

1S/C21H28Cl2N2/c1-5-12-9-14(18(22)16(7-3)20(12)24)11-15-10-13(6-2)21(25)17(8-4)19(15)23/h9-10H,5-8,11,24-25H2,1-4H3

InChI key

VIOMIGLBMQVNLY-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 88 °C
Melting range (upper value): ≤ 90 °C
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service