Skip to Content
Merck
All Photos(1)

Key Documents

289051

Sigma-Aldrich

Ethyl 2-quinoxalinecarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2O2
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

81-83 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)c1cnc2ccccc2n1

InChI

1S/C11H10N2O2/c1-2-15-11(14)10-7-12-8-5-3-4-6-9(8)13-10/h3-7H,2H2,1H3

InChI key

HZMKSGSOBKQGJX-UHFFFAOYSA-N

Application

Ethyl 2-quinoxalinecarboxylate may be used in chemical synthesis studies.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service