Z531111
Aldrich® coiled condenser
unjacketed, L 200 mm, joint: ST/NS 29/32
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
Productos recomendados
Junta
joint: ST/NS29/32
Longitud
200 mm
Descripción general
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)