Documentos clave
Z530468
Sigma-Aldrich
Aldrich® Claisen distillation head
Joints: ST/NS 24/40 (3), thermometer joint: ST/NS 10/30
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
thermometer joint: ST/NS10/30
Joints: ST/NS24/40 (3)
immersion depth
75 mm
Categorías relacionadas
General description
Claisen adapter with a thermometer joint. Male side-arm drip-tip joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)