Merck
  • Developing high-affinity decoy receptors to treat multiple myeloma and diffuse large B cell lymphoma.
Developing high-affinity decoy receptors to treat multiple myeloma and diffuse large B cell lymphoma.
The Journal of experimental medicine (2022-07-27)
Yu Rebecca Miao, Kaushik Thakkar, Can Cenik, Dadi Jiang, Kazue Mizuno, Chenjun Jia, Caiyun Grace Li, Hongjuan Zhao, Anh Diep, Yu Xu, Xin Eric Zhang, Teddy Tat Chi Yang, Michaela Liedtke, Parveen Abidi, Wing-Sze Leung, Albert C Koong, Amato J Giaccia
ABSTRACT
Disease relapse and treatment-induced immunotoxicity pose significant clinical challenges for patients with hematological cancers. Here, we reveal distinctive requirements for neutralizing TNF receptor ligands APRIL and BAFF and their receptor activity in MM and DLBCL, impacting protein translation and production in MM cells and modulating the translation efficiency of the ATM interactor (ATMIN/ACSIZ). Therapeutically, we investigated the use of BCMA decoy receptor (sBCMA-Fc) as an inhibitor of APRIL and BAFF. While wild-type sBCMA-Fc effectively blocked APRIL signaling in MM, it lacked activity in DLBCL due to its weak BAFF binding. To expand the therapeutic utility of sBCMA-Fc, we engineered an affinity-enhanced mutant sBCMA-Fc fusion molecule (sBCMA-Fc V3) 4- and 500-fold stronger in binding to APRIL and BAFF, respectively. The mutant sBCMA-Fc V3 clone significantly enhanced antitumor activity against both MM and DLBCL. Importantly, we also demonstrated an adequate toxicity profile and on-target mechanism of action in nonhuman primate studies.
Sigma-Aldrich
Anti-ASCIZ Antibody, from rabbit, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.