Skip to Content
Merck
All Photos(1)

Key Documents

SRP6287

Sigma-Aldrich

LIF human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CDF, DIA, HILDA, LIF, MLPLI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 19.9 kDa (with Gly-Pro at the N- terminus)
observed mol wt 35-45 kDa (DTT-reduced.Ser 23 is the predicted N-terminus .)

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LIF(3976)

General description

Leukemia inhibitory factor (LIF) is a secreted pleiotropic glycoprotein which is a member of the gp130 signaling of interleukin-6 (IL-6) family cytokines. This protein was originally identified as a protein that prevented the growth of a leukemic cell line.

Biochem/physiol Actions

Leukemia inhibitory factor (LIF) suppresses the differentiation of myoblasts, and inhibits the activation of caspase-3 and blocks the reduction in Bcl-xL (B-cell lymphoma-extra large) levels, thus, preventing doxorubicin-induced apoptosis. Along with transforming growth factor β2 (TGFβ2)/fibroblast growth factor 2 (FGF2), LIF regulates nephrogenesis. It facilitates tubular regeneration following acute kidney failure. It plays a crucial role in neuroprotective pathways induced by preconditioning-induced oxidative stress. LIF is capable of maintaining the totipotency of embryonic stem cells. It is also involved in controlling the differentiation of embryonic stem cells, hematopoietic and neuronal cells.

Physical form

Sterile filtered through a 0.22 micron filter. Lyophilized from 1 x PBS, pH 7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile deionized water to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective effects of leukemia inhibitory factor against oxidative stress during high glucose-induced apoptosis in podocytes.
Xu J et al
Cell Stress & Chaperones, 17(4), 485-493 (2012)
The gp130 receptor cytokine family: regulators of adipocyte development and function.
White UA and Stephens JM
Current Pharmaceutical Design, 17(4), 340-346 (2011)
Complex conserved organization of the mammalian leukemia inhibitory factor gene: regulated expression of intracellular and extracellular cytokines.
Haines BP et al
Journal of Immunology, 162(8), 4637-4646 (1999)
Leukemia inhibitory factor (LIF) and its biological activity.
Chodorowska G et al
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina, 59(2), 189-193 (2004)

Articles

Leukemia Inhibitory Factor (LIF) is a stem cell growth factor used for the in vitro culture of pluripotent mouse embryonic stem cells (ES cells).

Read article on hematopoietic cytokines and hematopoiesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service