Skip to Content
Merck
All Photos(2)

Documents

SRP0433

Sigma-Aldrich

PDE5A1 active murine

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

CN5N, phosphodiesterase 5A, cGMP-specific

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

124 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.34 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... PDE5A(242202)

General description

Mouse PDE5A (GenBank Accession No. NM_153422) full length with N-terminal GST-tag, MW=124 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anindita Das et al.
The Journal of biological chemistry, 280(13), 12944-12955 (2005-01-26)
We investigated the effect of sildenafil in protection against necrosis or apoptosis in cardiomyocytes. Adult mouse ventricular myocytes were treated with sildenafil (1 or 10 microM) for 1 h before 40 min of simulated ischemia (SI). Necrosis was determined by
Hunter C Champion et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(5), 1661-1666 (2005-01-26)
The molecular mechanism for priapism is not well characterized. Although the nitric oxide (NO) pathway is known to mediate penile erection under normal conditions, we hypothesized that the mechanism of priapism rests in aberrant downstream signaling of this pathway based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service