NIST2638A
Carbon monoxide in nitrogen
NIST SRM 2638a, nominal 5000 umol/mol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
Recommended Products
form
gas (compressed)
packaging
pkg of cylinder
manufacturer/tradename
NIST®
application(s)
air monitoring
environmental
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Certified for the analytes listed below.
Carbon monoxide (CO)
Reference/Informational value is provided for the following analytes.
Carbon Dioxide (CO2), Water (H2O), Methane (CH4), Total other hydrocarbons, Nitrogen (N)
See certificate for values and more details at nist.gov/SRM.
Carbon monoxide (CO)
Reference/Informational value is provided for the following analytes.
Carbon Dioxide (CO2), Water (H2O), Methane (CH4), Total other hydrocarbons, Nitrogen (N)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Liquefied gas - Repr. 1A
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service