95781
TOC standard
according to ISO/CEN EN1484, 250 mg/L±5 mg/L
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
Quality Level
grade
analytical standard
Agency
according to ISO/CEN EN1484
quality
stabilized
shelf life
limited shelf life, expiry date on the label
concentration
250 mg/L±5 mg/L
technique(s)
TOC: suitable
suitability
in accordance for application
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
single component solution
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Total organic carbon (TOC) is a measure of the total amount of carbon in an organic compound. It is a critical parameter to evaluate the water quality for possible organic contamination.
Application
This analytical standard is intended for the calibration and validation of analytical instruments used to measure the total organic carbon (TOC) of drinking water, groundwater, surface water, seawater, and wastewater in accordance with ISO/CEN EN1484.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service