GF58275883
Palladium-silver alloy, Pd77Ag23
foil, 50 x 50mm, 0.125mm thickness, as rolled
Synonym(s):
PD270250
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
foil (temper - as rolled)
manufacturer/tradename
Goodfellow 582-758-83
size × thickness
50 mm × 50 mm × 0.125 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service