PHL89257
α-Pinene
phyproof® Reference Substance
Synonym(s):
(±)-2-Pinene, 2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (GC)
form
liquid
autoignition temp.
491 °F
manufacturer/tradename
PhytoLab
refractive index
n20/D 1.465 (lit.)
bp
155-156 °C (lit.)
density
0.858 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
[H][C@@]12CC=C(C)[C@@]([H])(C1)C2(C)C
InChI
1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m1/s1
InChI key
GRWFGVWFFZKLTI-RKDXNWHRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service