Skip to Content
Merck
All Photos(1)

Key Documents

82017-U

Hamilton® PRP-X100 (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, PEEK hardware

Synonym(s):

PRP-X100 column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€997.00

€997.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€997.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

€997.00


Please contact Customer Service for Availability

Product Name

Hamilton PRP-X100 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm, PEEK hardware

material

PEEK column

manufacturer/tradename

Hamilton

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

ammonium ion, quaternary phase

particle size

5 μm

pore size

100 Å

operating pH

1-14

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

The Hamilton PRP-X100, polymeric HPLC columns used for the separation of anions in ion chromatography.

Application


  • Fluoride effects on cell viability and ENaC expression in kidney epithelial cells.: The research investigates the effects of fluoride on cell viability and ENaC expression in kidney epithelial cells, highlighting the broader implications of fluoride chemistry in biological systems. The study provides insights into fluoride′s impact at the cellular level, useful for analytical chemists studying biological interactions (Santesso MR et al., 2021).

  • TMT-based quantitative proteomics analysis reveals the response of tea plant (Camellia sinensis) to fluoride.: Utilizing TMT-based quantitative proteomics, this research details the proteomic response of tea plants to fluoride exposure, providing a comprehensive analysis beneficial for chemists involved in plant biochemistry and analytical chemistry (Liu Y et al., 2018).

  • Label-free electrochemical detection of the human adenovirus 40/41 fiber gene.: This article presents a label-free electrochemical method for detecting human adenovirus genes, employing fluoride-based electrochemical processes. The research showcases applications in virology and analytical detection techniques (Song MJ and Jin JH, 2015).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service