Skip to Content
Merck
All Photos(1)

Key Documents

3135-U

Supelco

Female Luer to ¼ in. Hose Barb (nylon)

pkg of 2 ea

Synonym(s):

Air Sampling Pump Accessories

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
€10.60

€10.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 EA
€10.60

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

€10.60


Please contact Customer Service for Availability

Request a Bulk Order

packaging

pkg of 2 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for connecting air sampling tubes with male luer to pump tubing

air sampling device

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service