TMO159934
Nunc® EasYFlasks™, Nunclon® Δ Surface
polystyrene, capacity 70 mL, Nunclon® Delta surface, sterile, case of 30 ea
Synonym(s):
cell culture flask, treated flask
All Photos(1)
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
Recommended Products
material
polystyrene
sterility
sterile
packaging
case of 30 ea
manufacturer/tradename
Nunc 159934
capacity
70 mL
General description
Nunc® EasYFlasks™, Nunclon® Δ Surface
- Nunclon® Δ certified: a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
- Ergonomic design with a 1/3 turn to open, close or vent
- Low profile to maximize incubator space
- Volume graduations printed and molded on both sides
- Slightly angled side walls for complete viewing under a microscope
- Sterilized by gamma irradiation and certified to be nonpyrogenic
- Nunclon Delta Certification, including testing on four different cell types, provided in each case
- Vent/close caps feature a “Y” mark that allows visual verification of vent position.
- Vent position easily distinguished even when flasks are stacked in incubators.
- Filter membrane provides effective barrier against micro-organisms.
- Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation.
- Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances.
Legal Information
EasYFlasks is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries