Skip to Content
Merck
All Photos(1)

Key Documents

SML0436

Sigma-Aldrich

BNS-22

≥98% (HPLC)

Synonym(s):

8-[(3,4-Dihydro-2H-quinolin-1-yl)carbonyl]-5,7-dimethoxy-4-propyl-2H-chromen-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€105.00
25 MG
€270.00

€105.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€105.00
25 MG
€270.00

About This Item

Empirical Formula (Hill Notation):
C24H25NO5
CAS Number:
Molecular Weight:
407.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€105.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution; warmed)

storage temp.

2-8°C

SMILES string

COC1=C2C(OC(C=C2CCC)=O)=C(C(N3C(C=CC=C4)=C4CCC3)=O)C(OC)=C1

InChI

1S/C24H25NO5/c1-4-8-16-13-20(26)30-23-21(16)18(28-2)14-19(29-3)22(23)24(27)25-12-7-10-15-9-5-6-11-17(15)25/h5-6,9,11,13-14H,4,7-8,10,12H2,1-3H3

InChI key

LRPUQCTZOSTSGL-UHFFFAOYSA-N

Biochem/physiol Actions

BNS-22 is a DNA-topoisomerase II (TOPO2) catalytic inhibitor.
BNS-22 is a derivative of natural product GUT-70 isolated from the stem bark of Calophyllum brasiliense exhibits antiproliferative activity against human cancer cells. It appears that BNS-22 is a specific DNA-topoisomerase II (TOPO2) catalytic inhibitor. BNS-22 does not induce DNA damage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service