EP0030128532
Eppendorf® Twin tec PCR plates, 384 wells
blue plate, conical bottom blue wells, pkg of 25 ea
Synonym(s):
PCR Plates
Select a Size
€228.00
Select a Size
About This Item
conical bottom blue wells
polypropylene
€228.00
Recommended Products
material
blue plate
conical bottom blue wells
polypropylene
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128532
technique(s)
PCR: suitable
size
384 wells
working volume
40 μL
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Standard PCR
- Real time PCR
- High throughput PCR
Features and Benefits
- “Eppendorf” Twin tec PCR plates have skirted wells with a maximum volume of 40?L.
- Enhanced rim design for efficient barcode scanning and automated workflows.
- The plates can be stacked together enabling convenient storage and handling.
- These Twin tec PCR plates are compatible with most 384- well theromcyclers.
- Well suited for quantitative real time PCR applications, featuring OptiTrack® matrix for quicker sample identification and lowering pipetting errors.
- Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service