Skip to Content
Merck
All Photos(2)

Key Documents

MABN2443

Sigma-Aldrich

Anti-nonphospho-GSK3 (Ser9) Antibody, beta specific Antibody, clone 12B2

clone 12B2, from mouse

Synonym(s):

Glycogen synthase kinase-3 beta, EC: 2.7.11.26, GSK-3 beta, Serine/threonine-protein kinase GSK3B

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
200 μL
€375.00

€153.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
200 μL
€375.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€153.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12B2, monoclonal

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer9)

Gene Information

human ... GSK3B(2932)

General description

Glycogen synthase kinase-3 beta (UniProt: P49841; also known as EC: 2.7.11.26, GSK-3 beta, Serine/threonine-protein kinase GSK3B) is encoded by the GSK3B gene (Gene ID: 2932) in human. GSK-3 is a multifunctional serine/threonine kinase, is a key regulator of numerous signaling pathways during embryogenesis and in metabolic control. Two isoforms of GSK-3 are reported in mammals: a 51 kDa GSK-3 alpha and a 47 kDa GSK-3 beta. The GSK-3 alpha contains a glycine-rich extension at its N-terminal. GSK-3 is constitutively active in cells and is regulated through inhibition of its activity. GSK-3 shows a preference for target proteins that are pre-phosphorylated at a `priming residue located C-terminal to the site of GSK-3 phosphorylation. Priming phosphorylation, although not absolutely required, enhances the efficiency of phosphorylation of most GSK-3 substrates. Phosphorylation of a threonine residue in the activation loop (T-loop) is considered to be essential for its activity. The T-loop of GSK-3 beta is phosphorylated at Tyr216, which play a role in forcing open the substrate-binding site of the enzyme. A primary mechanism of GSK-3 beta activity regulation is phosphorylation of N-terminal serine residues (Serine 9). Phosphorylation at this site is inhibitory to GSK-3 beta kinase activity because the phosphorylated N-terminus acts as a competitive inhibitor for primed substrates. GSK-3 beta is constitutively active in resting cells and treatment of cells with an agent, such as insulin, causes GSK-3 inactivation through a PI 3-kinase (PI 3-K)-dependent mechanism.

Specificity

Clone 12B2 detects GSK-3 beta in human, murine, and rat cells.

Immunogen

Epitope: N-terminus
KLH-conjugated three linear peptides from the N-terminal region of human GSK-3 beta.

Application

Anti-nonphospho-GSK3 (Ser9), beta specific, clone 12B2, Cat. No. MABN2443, is a highly specific mouse monoclonal antibody that targets Glycogen synthase kinase-3 beta and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected nonphospho-GSK3 (Ser9) in human and mouse brain tissue lysates.

ELISA Analysis: A representative lot detected nonphospho-GSK3 (Ser9) in ELISA applications (Grabinski, T., et. al. (2016). Front Mol Neurosci. 9:123).

Western Blotting Analysis: A representative lot detected nonphospho-GSK3 (Ser9) in Western Blotting applications (Grabinski, T., et. al. (2016). Front Mol Neurosci. 9:123).

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected nonphospho-GSK3 (Ser9) in human cerebral cortex tissue.

Immunocytochemistry Analysis: A representative lot detected nonphospho-GSK3 (Ser9) in Western Blotting applications (Grabinski, T., et. al. (2016). Front Mol Neurosci. 9:123).

Immunoprecipitation Analysis: A representative lot detected nonphospho-GSK3 (Ser9) in Western Blotting applications (Grabinski, T., et. al. (2016). Front Mol Neurosci. 9:123).

Immunohistochemistry Analysis: A representative lot detected nonphospho-GSK3 (Ser9) in Western Blotting applications (Grabinski, T., et. al. (2016). Front Mol Neurosci. 9:123).

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected nonphospho-GSK3 (Ser9) in rat brain tissue lysate.

Target description

~45 kDa observed. 46.74 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service