Skip to Content
Merck
All Photos(2)

Key Documents

MABE1818

Sigma-Aldrich

Anti-PP6C Antibody, clone 7G7

clone 7G7, from mouse

Synonym(s):

Serine/threonine-protein phosphatase 6 catalytic subunit, EC3.1.3.16

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€342.00

€130.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€342.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7G7, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PPP6C(5537)

General description

Serine/threonine-protein phosphatase 6 catalytic subunit (UniProt: O00743; also known as EC: 3.1.3.16, PP6C) is encoded by the PPP6C (also known as PPP6) gene (Gene ID: 5537) in human. Protein phosphatase 6 (PP6) is a serine/threonine protein phosphatase that is classified as a type 2A phosphatase family member based on its sequence homology to the catalytic subunit of PP2A. It is a component of a signaling pathway regulating cell cycle progression in response to IL2 receptor stimulation. The PP6 holoenzyme is a heterotrimeric complex of a catalytic subunit, a SAPS domain-containing subunit (PP6R) and an ankyrin repeat-domain containing regulatory subunit (ARS). PP6C is the catalytic subunit of PP6 that has multiple metal binding (manganese) sites and binds 2 manganese ions per subunit. It is ubiquitously expressed in all tissues and highest expression levels are observed in in testis, heart, kidney, brain, stomach, liver and skeletal muscle. PP6C is shown to be upregulated in the glioblastoma and its levels correlate with patient survival time. Silencing of PP6C by small interfering RNA induces sensitivity to IR and delayed release from the G2/M checkpoint. Three isoforms of PP6C have been described that are produced by alternative splicing. (Ref.: Shen, Y et al. (2011). Cell Death and Dis. 2, e241; Douglas, P., et al. (2010). Mol. Cell. Biol. 30(6); 1368-1381).

Specificity

Clone 7G7 specifically detects human Serine/threonine-protein phosphatase 6 catalytic subunit. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Serine/threonine-protein phosphatase 6 catalytic subunit.

Application

Anti-PP6C, clone 7G7, Cat. No. MABE1818, is a highly specific mouse monoclonal antibody that targets Serine/threonine-protein phosphatase 6 catalytic subunit and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected PP6C in lysate from NIH3T3 mouse fibroblasts ectopically expressing N-terminally tagged human PP6 catalytic subunit. (Courtesy of Dr. Stefan Schuechner and Dr. Egon Ogris, Medical University of Vienna, Austria).

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PP6C in MCF7 cell lysate.

Target description

~35 kDa observed; 35.14 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service