Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB199P

Sigma-Aldrich

Milli-Mark® Anti-CD56-PE Antibody, clone C5.9

clone C5.9, Milli-Mark®, from mouse

Synonym(s):

Neural cell adhesion molecule 1, NCAM-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€370.00

€370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€370.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C5.9, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NCAM1(4684)

Related Categories

General description

CD56 is a membrane glycoprotein which has multiple isoforms generated by alternative splicing from a single gene located on chromosome 11 (Costa, 1997). The core polypeptide is the 140 kDa isoform which can be variably glycosylated to produce mature species. The predominant isoform in NK and T cells is the transmembrane-anchored 140 kDa protein. In tissues, CD56 antigen is found in the cerebellum and cortex of the brain, and at neuromuscular junctions. This molecule is identical to the 140 kDa isoform of the neuronal cell adhesion molecule (N-CAM) (Poggi, 2002; Lanier, 1989). Additionally, CD56 is also present in certain large granular lymphocyte leukemias, small cell lung carcinomas, neural-derived tumors, myelomas, and myeloid leukemias (Poggi, 2002).

Specificity

Antibody recognizes Human CD56 antigen.

Immunogen

Human NK Cells

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD56-PE Antibody, clone C5.9 is validated for use in FC for the detection of CD56.

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

140 kDa (Poggi, 2002; Lanier, 1989)

Physical form

Protein A purified
Purified Mouse monoclonal IgG2b conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service