Skip to Content
Merck
All Photos(1)

Key Documents

AP506R

Sigma-Aldrich

Goat Anti-Human Ig λ chain Antibody, Rhodamine (TRITC) conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€278.00

€278.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€278.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€278.00


Please contact Customer Service for Availability

Request a Bulk Order

conjugate

rhodamine

Quality Level

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable

General description

Prepared from pooled antisera from goats hyperimmunized with human myeloma proteins with lambda light chains.

Specificity

The antibody reacts with human lambda light chains as demonstrated by ELISA and flow cytometry.

Application

Epitope: Ig lambda lightchain

Physical form

Purified by affinity chromatography on pooled human Igs with lambda light chains covalently linked to agarose. Liquid in 1.0 mL PBS/NaN3.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. Do not freeze.

Other Notes

Reacts with human lambda light chains. Absorbed for human myeloma proteins with kappa light chains.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service