Skip to Content
Merck
All Photos(1)

Key Documents

AP1173

Sigma-Aldrich

Anti-EPHA4 Mouse mAb (6H7)

liquid, clone 6H7, Calbiochem®

Synonym(s):

Anti-Ephrin Receptor A4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€366.00

€366.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€366.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€366.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6H7, monoclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPHA4(2043)

General description

Anti-EPHA4, mouse monoclonal, clone 6H7, recognizes the EPHA4 protein in NIH3T3 cells. It is validated for use in ELISA and Western blotting.
Purified mouse monoclonal antibody. Recognizes the ~100 kDa EPHA4 protein.
Recognizes the EPHA4 protein in NIH3T3 cells.

Immunogen

A recombinant polypeptide corresponding to amino acids of 887-987 human EPHA4, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Hitt et al.
The Journal of biological chemistry, 287(46), 38408-38425 (2012-09-19)
BACE1 is the β-secretase enzyme that initiates production of the β-amyloid peptide involved in Alzheimer disease. However, little is known about the functions of BACE1. BACE1-deficient mice exhibit mild but complex neurological phenotypes suggesting therapeutic BACE1 inhibition may not be
Heng-Huan Lee et al.
Nature communications, 12(1), 2788-2788 (2021-05-15)
Human ribonuclease 1 (hRNase 1) is critical to extracellular RNA clearance and innate immunity to achieve homeostasis and host defense; however, whether it plays a role in cancer remains elusive. Here, we demonstrate that hRNase 1, independently of its ribonucleolytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service