Skip to Content
Merck
All Photos(1)

Key Documents

8.55086

Sigma-Aldrich

HMBA-NovaGel

Novabiochem®

Synonym(s):

4-Hydroxymethylbenzoic acid NovaGel

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€81.90
25 G
€1,330.00

€81.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€81.90
25 G
€1,330.00

About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

€81.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

NovaGel
Novabiochem®

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-8°C

General description

This resin is prepared by derivatization of aminomethyl NovaGel with the base-labile 4-hydroxymethylbenzoic acid linker. NovaGel resins combine the high functionality of polystyrene resins with the excellent swelling properties of PEG-PS type supports [1].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] J. H. Adams, et al. (1998) J. Org. Chem., 63, 3706.

Linkage

Replaces: 01-64-0285

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service