Skip to Content
Merck
All Photos(1)

Key Documents

8.20991

Sigma-Aldrich

1,3-Phenylenediamine

for synthesis

Synonym(s):

1,3-Phenylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€27.90
250 G
€49.50
1 KG
€148.00

€27.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€27.90
250 G
€49.50
1 KG
€148.00

About This Item

Linear Formula:
1,3-(NH2)2C6H4
CAS Number:
Molecular Weight:
108.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-584-7
NACRES:
NA.22

€27.90


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.3 hPa ( 100 °C)

Quality Level

form

solid

autoignition temp.

560 °C

potency

280 mg/kg LD50, oral (Rat)

pH

8 (20 °C, 100 g/L in H2O)

bp

283-284 °C/1013 hPa

mp

63-64 °C

transition temp

flash point 187 °C

solubility

350 g/L

density

1.14 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

Nc1cc(ccc1)N

InChI

1S/C6H8N2/c7-5-2-1-3-6(8)4-5/h1-4H,7-8H2

InChI key

WZCQRUWWHSTZEM-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 62 °C
Melting range (upper value): ≤ 64 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service