Skip to Content
Merck
All Photos(1)

Key Documents

1.14963

Supelco

Crack Set 20 Pretreatment Kit

sufficient for 90 digestions of nitrogen (total), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€168.00

€168.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
1 SET
€168.00

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

€168.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Crack Set 20, sufficient for 90 digestions of nitrogen (total), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 90 digestions of nitrogen (total)

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

storage temp.

15-25°C

General description

for the digestion of nitrogen (total) 90 digestions Spectroquant®

Analysis Note

Application test: conforms

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service