Skip to Content
Merck

1.00250

SAFC

Phosphoric acid 75%

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Orthophosphoric acid, o-Phosphoric acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 L
€199.00
200 L
Inquire
950 L
Inquire

€199.00


Estimated to ship onApril 03, 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
10 L
€199.00
200 L
Inquire
950 L
Inquire

About This Item

UNSPSC Code:
12352301

€199.00


Estimated to ship onApril 03, 2025Details


Request a Bulk OrderRequest a Sample

vapor pressure

2 hPa ( 20 °C)

Quality Level

product line

EMPROVE® EXPERT

pH

<0.5 (20 °C, 100 g/L in H2O)

kinematic viscosity

14.7 cSt(25 °C)

bp

135 °C

mp

-17 °C

density

1.58 g/cm3 at 20 °C

application(s)

parenterals

storage temp.

2-8°C

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service