Skip to Content
Merck
All Photos(1)

Key Documents

900143J

Avanti

PGB2

Avanti Research - A Croda Brand

Synonym(s):

15-Hydroxy-9-oxoprosta-5,8(12),13-trienoic acid, 9-oxo-15S-hydroxy-prosta-5Z,8(12),13E-trien-1-oic acid, Prostaglandin B2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic

Assay

>99% (TLC)

form

methyl acetate solution

packaging

pkg of 1X5 mg (900143J-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\C1=C(C\C=C/CCCC(O)=O)C(=O)CC1

InChI

1S/C20H30O4/c1-2-3-6-9-17(21)14-12-16-13-15-19(22)18(16)10-7-4-5-8-11-20(23)24/h4,7,12,14,17,21H,2-3,5-6,8-11,13,15H2,1H3,(H,23,24)/b7-4-,14-12+/t17-/m0/s1

InChI key

PRFXRIUZNKLRHM-HKVRTXJWSA-N

Application

Lipids research

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service