Key Documents
Z277789
Sigma-Aldrich
HGS Researcher model set
Inorganic chemistry-D
Select a Size
1 EA
€145.00
€145.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€145.00
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
€145.00
Please contact Customer Service for Availability
feature
no. of atoms 116
no. of bonds 180
manufacturer/tradename
HGS 1006
General description
Contain a variety of polyhedrons and bonds to meet the broad demand of chemical research. Scale: 1Å = 2.5 cm.
Researcher organic set is suited for building fundamental organic compounds, polymers, steroids, and some simple complex salts.
Researcher inorganic set is suited for building complicated tetra-, hexa-, and octa-coordinated complex compounds and chelate compounds. The fundamental structure of ionic crystals such as rock salt, cesium chloride, etc. can be constructed without supplementary parts.
Crystal structure set permits assembly of nearly all the fundamental crystal structures in inorganic chemistry and crystallography, as well as inorganic giant molecules.
Choose from one of the most recent versions:
Certificates of Analysis (COA)