Key Documents
Z225142
Sigma-Aldrich
Scienceware® long-handled scraper
reaches deep inside glass vessels without scratching
Select a Size
1 EA
€214.00
€214.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€214.00
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
€214.00
Please contact Customer Service for Availability
manufacturer/tradename
Bel-Art F368290001
L
46 1/4 in.
General description
Reaches deep inside glass vessels without scratching. The rigid nylon scraper blade (7 in. L × 4 1/2 in. W) is molded to a tubular, fiberglass shaft having a ribbed hand grip.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. How long of a length is Z225142 scraper ? Looking for similar at 20-30 inches and autoclavable ideally
    1 answer
    1. The total length of this scraper is 46.25 in. This includes the blade which is 7 in. long and 4.5 in. wide. This is a nylon product that is stable up to 100°C, but not suitable for autoclaving at 121°C. Other long-handle scapers available are comprised of HDPE and LDPE which are also not autoclavable.
      Helpful?