Key Documents
Z188832
Supelco
Black plastic caps
Solid top, PTFE-faced rubber liner, thread for 18-400, pkg of 200 ea
Select a Size
200 EA
€396.00
€396.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
200 EA
€396.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
€396.00
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE-faced rubber liner
black phenolic cap
feature
closure type black phenolic solid top cap
packaging
pkg of 200 ea
fitting
thread for 18-400
General description
These autoclavable caps are designed to ensure the ultimate in product safety.
Application
  • Marine litter accumulation along the Bulgarian Black Sea coast: Categories and predominance.: This research categorizes and quantifies the types of marine litter found along the Bulgarian Black Sea coast, with a particular focus on the prevalence of black plastic caps. The findings underscore the necessity for targeted clean-up efforts and policy interventions to address specific types of marine debris (Simeonova & Chuturkova, 2019).
Choose from one of the most recent versions:
Certificates of Analysis (COA)