Key Documents
Z125199
Ace Claisen Adapter
Standard
Synonym(s):
distilling adapter, glass adapter, glass adapters
Select a Size
1 EA
€95.00
€95.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€95.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€95.00
Please contact Customer Service for Availability
label
A
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 510154
Related Categories
General description
Threaded nylon bushing for 7 mm O.D. thermometer.
product is shown on the left in the photo
Choose from one of the most recent versions:
Certificates of Analysis (COA)