Skip to Content
Merck
All Photos(2)

Key Documents

GF63430949

Iridium

wire, 0.25 mm diameter, size 1000 mm, purity 99.9%

Synonym(s):

IR005121

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ir
CAS Number:
Molecular Weight:
192.22
MDL number:
UNSPSC Code:
12141720
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 634-309-49

resistivity

4.71 μΩ-cm

size

1000 mm

diameter

0.25 mm

bp

4130 °C (lit.)

mp

2450 °C (lit.)

density

22.65 g/cm3 (lit.)

SMILES string

[Ir]

InChI

1S/Ir

InChI key

GKOZUEZYRPOHIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Application

  • Iridium-based double perovskites for efficient water oxidation in acid media: This study discusses the potential of iridium-based double perovskites in enhancing the efficiency of water oxidation, crucial for energy storage and conversion technologies (O Diaz-Morales, S Raaijman, R Kortlever, 2016).
  • Balancing activity, stability and conductivity of nanoporous core-shell iridium/iridium oxide oxygen evolution catalysts: This research explores the design of nanoporous iridium-iridium oxide structures that optimize the activity and stability of catalysts used for oxygen evolution (YT Kim, PP Lopes, SA Park, AY Lee, J Lim, 2017).
  • Perspectives on current and future iridium demand and iridium oxide catalysts for PEM water electrolysis: Analyzing the demand projections for iridium in polymer electrolyte membrane water electrolysis, this paper assesses strategies to improve the utilization of iridium in catalysts (M Clapp, CM Zalitis, M Ryan, 2023).

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service