Skip to Content
Merck
All Photos(1)

Key Documents

BMSZ326BKR

Benchmark BucketBuddy Magnetic Basket and Tube Rack for Laboratory Centrfuges

Synonym(s):

centrifuge storage bucket

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€249.00

€249.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€249.00

About This Item

UNSPSC Code:
41122804
NACRES:
NB.13

€249.00


Please contact Customer Service for Availability

feature

to hold most capped tubes ranging from 1.5 ml to 50 ml

manufacturer/tradename

Benchmark Scientific Z326-BKR

General description

The BucketBuddy Magnetic Bucket and Tube Rack for Laborator Centrifuges, is the perfect companion to your Hermle centrifuge. The magnetic feature allows the user to place the BucketBuddy wherever they like on the magnetic surfaces of their centrifuge. This provides convenient storage, and clears up valuable benchspace and clutter around your centriuge. The BucketBuddy holds all Hermle buckets/inserts up to 750ml, and most capped tubes ranging from 1.5ml to 50ml.

Application

Centrifugation accessory, accessory storage, sample storage

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service