MilliporeSigma
HomeProtein PurificationSeparation Options with Superdex®
Separation Options with Superdex®
Superdex® is produced with different average particle sizes and different selectivities (see Table 2.2).
  • Use the prepacked columns with Superdex® Peptide, Superdex® 75, and Superdex® 200 Increase for small-scale preparative purification and analytical runs.
  • Use columns prepacked with Superdex® 30 prep grade, Superdex® 75 prep grade, or Superdex® 200 prep grade media for preparative applications.
  • Use bulk media of Superdex® 30 prep grade, Superdex® 75 prep grade, and Superdex® 200 prep grade for preparative purification with larger sample volumes of up to 13 mL.
  • The maximum pressure drop over the packed bed is a typical value. Note that the actual value is individual for each column and needs to be determined (see Setting column pressure limits, Chapter 1).
  • Start with Superdex® 200 Increase when the molecular weight of the protein of interest is unknown. Superdex® 200 Increase or Superdex® 200 prep grade are especially suitable for the separation of monoclonal antibodies from dimers and from contaminants of lower molecular weight, for example albumin and transferrin.
  • Start with Superdex® Peptide or Superdex® 30 prep grade for separations of peptides, oligonucleotides and small proteins below Mr 10 000.
  • Exposure to temperatures outside the range 4 °C to 40 °C will negatively affect the efficiency of the packed bed and the column will need to be repacked.
Product*Fractionation range, Mr (globular proteins)Sample Loading capacity†Maximum pressure drop over the packed bedRecommended Operational Flow rate‡
Superdex® 200 Increase 3.2/3001 × 104 to 6 × 1054 to 50 μL3.0 MPa, 30 bar, 435 psi**0.075 mL/min
Superdex® 200 Increase 5/150 GL1 × 104 to 6 × 1054 to 50 μL3.0 MPa, 30 bar, 435 psi**0.45 mL/min
Superdex® 200 Increase 10/300 GL1 × 104 to 6 × 10525 to 500 μL3.0 MPa, 30 bar, 435 psi**0.075 mL/min
Superdex® Peptide 3.2/3001 × 102 to 7 × 103< 50 μL2.0 MPa, 20 bar, 290 psi0.05 mL/min
Superdex® Peptide 10/300 GL1 × 102 to 7 × 10325 to 500 μL2.0 MPa, 20 bar, 290 psi< 1.0 mL/min
HiLoad 16/600 Superdex® 30 pg§< 1 × 104≤ 5 mL3 MPa, 3 bar, 42 psi1.0 mL/min
HiLoad 26/600 Superdex® 30 pg§< 1 × 104≤ 13 mL3 MPa, 3 bar, 42 psi2.6 mL/min
Superdex® 30 pg§ (Bulk medium)< 1 × 1040.5% to 4% of total column volumeColumn-dependent10 to 50 cm/h
Superdex® 75 3.2/3003 × 103 to 7 × 104< 50 μL2.4 MPa, 24 bar, 350 psi0.05 mL/min
Superdex® 75 5/150 GL3 × 103 to 7 × 1044 to 50 μL1.8 MPa, 18 bar, 260 psi0.3 mL/min
Superdex® 75 10/300 GL3 × 103 to 7 × 10425 to 500 μL1.8 MPa, 18 bar, 260 psi0.5 to 0.75 mL/min
HiLoad 16/600 Superdex® 75 pg§3 × 103 to 7 × 104≤ 5 mL0.3 MPa, 3 bar, 42 psi1.0 mL/min
HiLoad 26/600 Superdex® 75 pg§3 × 103 to 7 × 104≤ 13 mL0.3 MPa, 3 bar, 42 psi2.6 mL/min
Superdex® 75 pg§ (Bulk medium)3 × 103 to 7 × 1040.5% to 4% of total column volumeColumn-dependent10 to 50 cm/h
Superdex® 200 3.2/3001 × 104 to 6 × 105< 50 μL1.5 MPa, 15 bar, 220 psi0.05 mL/min
Superdex® 200 5/150 GL1 × 104 to 6 × 1054 to 50 μL1.5 MPa, 15 bar, 220 psi0.3 mL/min
Superdex® 200 10/300 GL1 × 104 to 6 × 10525 to 500 μL1.5 MPa, 15 bar, 220 psi0.5 to 0.75 mL/min
HiLoad 16/600 Superdex® 200 pg§1 × 104 to 6 × 105≤ 5 mL0.3 MPa, 3 bar, 42 psi1.0 mL/min
HiLoad 26/600 Superdex® 200 pg§1 × 104 to 6 × 105≤ 13 mL0.3 MPa, 3 bar, 42 psi2.6 mL/min
Superdex® 200 pg§ (Bulk medium)1 × 104 to 6 × 1050.5% to 4% of total column volumeColumn-dependent10 to 50 cm/h
Table 2.2.Separation options with Superdex® media
* HiLoad is packed with Superdex® prep grade.
† For maximum resolution, apply as small sample volume as possible. Note that sample volumes less than 0.5% normally do not improve resolution.
‡ See Appendix 5 to convert linear flow (cm/h) to volumetric flow rates (mL/min) and vice versa. § prep grade.
** Typical pressure drop over the packed bed.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch