MilliporeSigma
HomeProtein PurificationCharacteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (High Sub) Media and Columns
Characteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (High Sub) Media and Columns
Glutathione Sepharose High Performance is recommended for high-resolution purification of GST-tagged proteins, providing sharp peaks and concentrated eluent. Glutathione Sepharose 4 Fast Flow is excellent for scaling up. Glutathione Sepharose 4B has high capacity and is recommended for packing small columns and other formats including batch purifications.
Table A5.1 summarizes key characteristics of these three Glutathione Sepharose media, and Tables A5.2 to A5.6 summarize the characteristics of these media prepacked in columns and in 96-well filter plates. Table A5.7 summarizes key characteristics of HiTrap Benzamidine Fast Flow (high sub).
CharacteristicsGlutathione Sepharose High PerformanceGlutathione Sepharose 4 Fast FlowGlutathione Sepharose 4B
MatrixHighly cross-linked 6% agaroseHighly cross-linked 4% agarose4% agarose
Average particle size34 µm90 µm90 µm
Ligand concentration1.5–3.5 mg glutathione
/ml medium (based on Gly)
120–320 µmol glutathione/ml medium200–400 µmol glutathione/g washed and dried medium
Binding capacity17 mg recombinant GST/ml medium10 mg recombinant GST/ml medium25 mg horse liver GST/ml medium
Recommended flow velocity2< 150 cm/h50–300 cm/h< 75 cm/h
Chemical stabilityStable to all commonly used aqueous buffers, e.g., 1 M acetate, pH 4.0 and 6 M Gua-HCl for 1 h at room temperatureStable to all commonly used aqueous buffers, e.g., 1 M acetate, pH 4.0, and 6 M Gua-HCl for 1 h at room temperatureStable to all commonly used aqueous buffers, e.g., 0.1 M NaOH, 70% ethanol, or 6 M Gua-HCl for 2 h at room temperature or exposure to 1% (w/v) SDS for 14 d.
pH stability3–123–124–13
Storage temperature4 °C to 30 °C4 °C to 30 °C4 °C to 30 °C
Storage buffer20% ethanol20% ethanol20% ethanol
Table A5.1Characteristics of Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, and Glutathione Sepharose 4B
1 The binding of GST-tagged proteins depends on size, conformation, and concentration of the protein in the sample loaded. Binding of GST to glutathione is also flow dependent, and lower flow rates often increase the binding capacity. This is important during sample loading. Protein characteristics, pH, temperature, and the media used may also affect the binding capacity.
2 When using water at room temperature.
Chromatography mediaGST MultiTrap FF: Glutathione Sepharose 4 Fast Flow
GST MultiTrap 4B: Glutathione Sepharose 4B
Filter plate size1127.8 × 85.5 × 30.6 mm
Filter plate materialPolypropylene and polyethylene
Binding capacityGST MultiTrap FF: Up to 0.5 mg GST-tagged protein/well
GST MultiTrap 4B: Up to 0.5 mg GST-tagged protein/well
Reproducibility between wells2+/- 10%
Volume packed medium/well50 µl (500 µl of 10% slurry)
Number of wells96
Centrifugation speed:
Recommended
Maximum
Depends on sample pretreatment and sample properties
100–500 × g
700 × g
Vacuum pressure:
Recommended
Maximum
Depends on sample pretreatment and sample properties
-0.1 to -0.3 bar
-0.5 bar
pH stabilityGlutathione Sepharose 4 Fast Flow: 3–12
Glutathione Sepharose 4B: 4–13
Storage20% ethanol
Storage temperature4 °C to 30 °C
A5.2Characteristics of GST MultiTrap FF and GST MultiTrap 4B
1 According to American National Standards Institute (ANSI) and Society for Biomolecular Screening (SBS).
1-2004, 3-2004, and 4-2004 standards.
2 The amount of eluted target proteins/well does not differ more than +/- 10% from the average amount/well for the entire filter plate.
CharacteristicsGSTrap HPGSTrap FFGSTrap 4B
Chromatography mediaGlutathione Sepharose High PerformanceGlutathione Sepharose 4 Fast FlowGlutathione Sepharose 4B
Average particle size34 µm90 µm90 µm
Dynamic binding capacity1,2Approx. 7 mg rGST/ml mediumApprox. 10 mg rGST/ml mediumApprox. 25 mg horse liver GST/ml medium
Max. back pressure30.3 MPa, 3 bar0.3 MPa, 3 bar0.3 MPa, 3 bar
Recommended flow rate3Sample loading: 0.2–1 ml/min (1 ml) and 1–5 ml (5 ml)
Washing and elution:
1–2 ml/min (1 ml column) and 5–10 ml/min (5 ml column)
Sample loading: 0.2–1 ml/min (1 ml) and 1–5 ml (5 ml)
Washing and elution:
1–2 ml/min (1 ml column) and 5–10 ml/min (5 ml column)
Sample loading: 0.2–1 ml/min (1 ml) and 0.5–5 ml/min (5 ml)
Washing and elution:
1 ml/min (1 ml column) and 5 ml/min (5 ml column)
Chemical stabilityStable to all commonly used aqueous buffers, e.g. 1 M acetate, pH 4.0 and 6 M Gua-HCl for 1 h at room temperatureStable to all commonly used aqueous buffers, e.g. 1 M acetate, pH 6.0, and 6 M Gua-HCl for 1 h at room temperatureStable to all commonly used aqueous buffers. Exposure to 0.1 M NaOH, 70% ethanol, or 6 M Gua-HCl for 2 h at room temperature or to 1% (w/v) SDS for 14 d causes no significant loss of activity.
pH stability3–123–124–13
Storage temperature4 °C to 30 °C4 °C to 30 °C4 °C to 30 °C
Storage buffer20% ethanol20% ethanol20% ethanol
Table A5.3Characteristics of prepacked GSTrap HP, GSTrap FF, and GSTrap 4B columns
1 The binding of GST-tagged proteins depends on size, conformation, and concentration of the protein in the sample loaded. Binding of GST to glutathione is also flow dependent, and lower flow rates often increase the binding capacity. This is important during sample loading. Protein characteristics, pH, temperature, and the media used may also affect the binding capacity.
2 Dynamic binding capacity conditions (60% breakthrough):
Sample: 1 mg/ml pure GST-tagged protein in binding buffer
Column volume: 0.4 ml
Flow rate: 0.2 ml/min (60 cm/h)
Binding buffer: 10 mM sodium phosphate, 140 mM NaCl, 2.7 mM KCl, pH 7.4
Elution buffer: 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0
3 When using water at room temperature.
Chromatography mediumGlutathione Sepharose 4 Fast Flow
Column volume20 ml
Column dimensions1.6 × 10 cm
Dynamic binding capacity1,2Approx. 200 mg rGST/column
Recommended flow rate31–10 ml/min (30–300 cm/h)
Max. flow rate310 ml/min (300 cm/h)
Max. pressure over the packed bed during operation31.5 bar (0.15 MPa, 22 psi)
Column hardware pressure limit5 bar (0.5 MPa, 73 psi)
Storage0% ethanol
Storage temperature4 °C to 30 °C
A5.4Characteristics of GSTPrep FF 16/10
1 The binding of GST-tagged proteins depends on size, conformation, and concentration of the protein in the sample loaded. Binding of GST to glutathione is also flow dependent, and lower flow rates often increase the binding capacity. This is important during sample loading. Protein characteristics, pH, temperature, and the media used may also affect the binding capacity.
2 Dynamic binding capacity conditions (60% breakthrough):
Sample: 1 mg/ml pure GST-tagged protein in binding buffer
Column volume: 0.4 ml
Flow rate: 0.2 ml/min (60 cm/h)
Binding buffer: 10 mM sodium phosphate, 140 mM NaCl, 2.7 mM KCl, pH 7.4
Elution buffer: 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0
3 When using water at room temperature.
Column material fritsPolypropylene barrel, polyethylene
Column volume13 ml
MediumGlutathione Sepharose 4B
Average bead size90 µm
LigandGlutathione and 10-carbon linker arm
Ligand concentration7–15 µmol glutathione/ml medium
Protein binding capacity1Approx. 50 mg horse liver GST/column
Bed volume2 ml
Compatibility during useAll commonly used aqueous buffers
Chemical stabilityNo significant loss of the capacity is detected when Glutathione Sepharose 4B is exposed to 0.1 M citrate (pH 4.0), 0.1 M NaOH, 70% ethanol, or 6 M Gua-HCl2 for 2 h at room temperature. No significant loss of binding capacity is observed after exposure to 1% SDS for 14 d.
Storage solution20% ethanol
pH stability
4–13
Storage temperature4 °C to 30 °C
Table A5.5Characteristics of GST GraviTrap prepacked columns
Note: It is not recommended to autoclave the columns.
1 Binding capacity is protein dependent. The binding of GST-tagged proteins depends on size, conformation, and concentration of the protein in the sample. Binding of GST to glutathione is also flow dependent, and lower flow rates often increase the binding capacity. This is important during sample loading. Protein characteristics, pH, and temperature may also affect the binding capacity.
2 Exposing the sample to 6 M Gua-HCl will denature the GST tag. It is therefore important to remove all Gua-HCl before use.
Column material fritsPolypropylene barrel, polyethylene frits
Column volume900 µl
MediumGlutathione Sepharose 4B
Average bead size90 µm
LigandGlutathione and 10-carbon linker arm
Ligand concentration7–15 µmol glutathione/ml medium
Protein binding capacity1Approx. 500 mg horse liver GST/column
Bed volume50 µl
Compatibility during useAll commonly used aqueous buffers
Chemical stabilityNo significant loss of the capacity is detected when Glutathione Sepharose 4B is exposed to 0.1 M citrate (pH 4.0), 0.1 M NaOH, 70% ethanol or 6 M Gua-HCl2 for 2 h at room temperature. No significant loss of binding capacity is observed after exposure to 1% SDS for 14 d.
Storage solutionPBS and 0.05% Kathon™ CG/ICP Biocide
pH stability
4–13
Storage temperature4 °C to 30 °C
Table A5.6Characteristics of GST SpinTrap columns
1 Binding capacity is protein dependent. The binding of GST-tagged proteins depends on size, conformation, and concentration of the protein in the sample loaded. Binding of GST to glutathione is also flow dependent, and lower flow rates often increase the binding capacity. This is important during sample loading. Protein characteristics, pH, and temperature may also affect the binding capacity.
2 Exposing the sample to 6 M Gua-HCl will denature the GST tag. It is therefore important to remove all Gua-HCl before use.
Column dimensions (i.d. × h)0.7 × 2.5 cm (1 ml column) and 1.6 × 2.5 cm (5 ml column)
Column volumes1 ml and 5 ml
1 ml and 5 mlp-Aminobenzamidine (pABA)
Spacer14-atom
Ligand concentration12 µmol p-Aminobenzamidine/ml medium
Binding capacity35 mg trypsin/ml medium
Average particle size90 µm
Bead structureHighly cross-linked agarose, 4%
Maximum back pressure0.3 MPa, 3 bar
Recommended flow rates1 ml/min (1 ml column) and 5 ml/min (5 ml column)
Maximum flow rates4 ml/min (1 ml column) and 20 ml/min (5 ml column)
Chemical stabilityAll commonly used aqueous buffers
pH stability short term1pH 1–9
pH stability long term1pH 2–8
Storage temperature4 °C to 8 °C
Storage buffer20% ethanol in 0.05 M acetate buffer, pH 4
A5.7 Characteristics of HiTrap Benzamidine FF (high sub)
1 The ranges given are estimates based on our knowledge and experience. Please note the following: pH stability, short term refers to the pH interval for regeneration, cleaning-in-place, and sanitization procedures. pH stability, long term refers to the pH interval where the medium is stable over a long period of time without adverse effects on its chromatographic performance.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch