Protease Inhibitor Cocktails
Our complete collection of Protease Inhibitor Cocktails offers a diverse portfolio that accommodates your different sample needs. Specialized cocktails are available for various sample forms:
- Bacterial cells
- Mammalian tissues and cells
- Plant tissues and cells
- Histdine-tagged (HIS-tagged) proteins.
Our portfolio also includes protease inhibitor cocktails that are tailored to specific applications:
- We offer cocktails for customers who require animal component-free (ACF) protease inhibitor cocktails in their work.
- We also now offer the only fully mass spectrometry-compatible protease inhibitor cocktail and phosphatase inhibitor cocktail on the market, MSSAFE.
Our Protease Inhibitor Cocktails have been optimized to maintain and preserve protein functionality following cell lysis, and are available in powder, solution, or tablet forms. Component information for our cocktails is available in the table below:
Protease Inhibitor Cocktails | |||||
---|---|---|---|---|---|
Product Number | P2714 | S8820 | P1860 | P8465 | P8340 |
General Use | General Use | Tissue Culture | Bacterial Cells | Mammalian Tissues | |
Lyophilized powder - Water Soluble | Tablet - Water Soluble | Solution in DMSO | Lyophilized powder - Vial of DMSO | Solution in DMSO | |
AEBSF | ✓ | ✓ | ✓ | ✓ | ✓ |
Aprotinin | ✓ | ✓ | ✓ | ✓ | |
Bestatin | ✓ | ✓ | ✓ | ✓ | ✓ |
EDTA | ✓ | ✓ | ✓ | ||
E-64 | ✓ | ✓ | ✓ | ✓ | ✓ |
Leupeptin | ✓ | ✓ | ✓ | ✓ | |
Pepstatin A | ✓ | ✓ | ✓ | ||
Phosphoramidon | |||||
1,10-Phenanthroline | |||||
Elastinal | |||||
Nafamostat mesylate | |||||
Antipain |
Product Number | P8849 | S8830 | P8215 | P9599 | I3911 | I3786 | MSSAFE |
---|---|---|---|---|---|---|---|
Histidine- Tagged Proteins | Histidine- Tagged Proteins | Fungal/ Yeast Cells | Plant Cells | General Use (Animal Component Free/ACF) | Mammalian Cells and Tissues (Animal Component Free/ACF) | Mammalian Cells and Tissues | |
Solution in DMSO | Tablet - Water Soluble | Solution in DMSO | Solution in DMSO | Lyophilized powder (water soluble) | Solution in DMSO | Lyophilized powder (water soluble) | |
AEBSF | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | |
Aprotinin | ✓ | ✓ | ✓ | ||||
Bestatin | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | |
EDTA | ✓ | ||||||
E-64 | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | |
Leupeptin | ✓ | ✓ | ✓ | ✓ | |||
Pepstatin A | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | |
Phosphoramidon | ✓ | ✓ | ✓ | ||||
1,10-Phenanthroline | ✓ | ✓ | |||||
Elastinal | ✓ | ||||||
Nafamostat mesylate | ✓ | ||||||
Antipain | ✓ |
Additional Resource