Orchid Culture Media Formulations
Orchid tissue culture has its own unique requirements. Below is formulation information for a number of media that are used to successfully culture Orchidaceae.
K4003 - Knudson C modified orchid medium
P0931 - Phytamax orchid maintenance medium without charcoal
P1056 - Phytamax orchid medium with charcoal and banana powder
P6668 - Phytamax orchid maintenance medium
P6793 - Phytamax orchid multiplication medium
M5524 - Murashige and Skoog basal salt mixture (MS)
Component (mg/L) | K4003 | P0931 | P1056 | P6668 | P6793 | M5524* |
---|---|---|---|---|---|---|
Aluminum chloride • 6H2O | ||||||
Ammonium nitrate | 825.0 | 825.0 | 825.0 | 825.0 | 1650.0 | |
Ammonium phosphate monobasic | ||||||
Ammonium sulfate | 500.0 | |||||
Boric acid | 0.056 | 3.1 | 3.1 | 3.1 | 3.1 | 6.2 |
Calcium chloride anhydrous | 166.0 | 166.0 | 166.0 | 166.0 | 332.2 | |
Calcium nitrate | 694.4 | |||||
Calcium phosphate tribasic | ||||||
Cobalt chloride • 6H2O | 0.0125 | 0.0125 | 0.0125 | 0.0125 | 0.025 | |
Cupric sulfate • 5H2O | 0.0624 | 0.0125 | 0.0125 | 0.0125 | 0.0125 | 0.025 |
Na2-EDTA | 37.24 | 37.24 | 37.24 | 37.24 | 37.26 | |
ZnNa2-EDTA | ||||||
Ferric chloride • 6H2O | ||||||
Ferric citrate | ||||||
Ferric sulfate | ||||||
Ferric tartrate • 2H2O | ||||||
Ferrous sulfate • 7H2O | 25.0 | 27.85 | 27.85 | 27.85 | 27.85 | 27.8 |
Magnesium sulfate | 122.125 | 90.35 | 90.35 | 90.35 | 90.35 | 180.7 |
Manganese chloride • 4H2O | ||||||
Manganese sulfate • H2O | 5.682 | 8.45 | 8.45 | 8.45 | 8.45 | 16.9 |
Molybdenum trioxide | 0.016 | |||||
Molybdic acid (sodium salt) • 2H2O | 0.125 | 0.125 | 0.125 | 0.125 | 0.25 | |
Nickel chloride • 6H2O | ||||||
Nickel sulfate • 6H2O | ||||||
Potassium chloride | ||||||
Potassium iodide | 0.415 | 0.415 | 0.415 | 0.415 | 0.83 | |
Potassium nitrate | 950.0 | 950.0 | 950.0 | 950.0 | 1900.0 | |
Potassium phosphate monobasic | 250.0 | 85.0 | 85.0 | 85.0 | 85.0 | 170.0 |
Potassium sulfate | ||||||
Sodium nitrate | ||||||
Sodium phosphate monobasic | ||||||
Sodium sulfate | ||||||
Zinc nitrate • 6H2O | ||||||
Zinc sulfate • 7H2O | 0.331 | 5.3 | 5.3 | 5.3 | 5.3 | 8.6 |
Organics (mg/L) | K4003 | P0931 | P1056 | P6668 | P6793 | M5524* |
---|---|---|---|---|---|---|
Activated charcoal | 2000.0 | 2000.0 | ||||
Adenine hemisulfate | ||||||
Agar | ||||||
Banana powder (50% malto-dextrin) | 30,000 | |||||
6-Benzylaminopurine (BA) | 2.0 | |||||
D-Biotin | ||||||
6gamma-gamma- Dimethylallylaminopurine | ||||||
Glycine (free base) | ||||||
Indole-3-acetic acid | ||||||
Indole-3-butyric acid | ||||||
Kinetin | ||||||
MES (free acid) | 1000.0 | 1000.0 | 1000.0 | 1000.0 | ||
myo-Inositol | 100.0 | 100.0 | 100.0 | 100.0 | ||
alpha-Naphthaleneacetic acid | 0.5 | |||||
Nictotinic acid (free acid) | 1.0 | 1.0 | 1.0 | 0.5 | ||
Peptone | 2000.0 | 2000.0 | 2000.0 | 2000.0 | ||
Pyridoxine • HCl | 1.0 | 1.0 | 1.0 | 0.5 | ||
Sucrose | 20,000 | 20,000 | 20,000 | 20,000 | 20,000 | |
Thiamine • HCl | 10.0 | 10.0 | 10.0 | 1.0 |
*Composition for M5524, Murashige and Skoog Basal Salt Mixture, listed for purposes of comparison only.