MilliporeSigma
HomeGas Chromatography (GC)GC Analysis of a 37-Component FAME Mix on Omegawax®
GC Analysis of a 37-Component FAME Mix on Omegawax®
GC Analysis of a 37-Component FAME Mix on Omegawax®
Conditions
ColumnOmegawax 250, 30 m x 0.25 mm I.D., 0.25 µm (24136)
Oven50 °C, 3.0 °C/min to 280 °C
Carrier Gashelium, 40 cm/sec constant
SampleSupelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
Injection1 µL50:1 split
Inj. Temp.240 °C
DetectorFID, 280 °C
Description
CategoriesAnalytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured IndustryAgriculture
Clinical
Food and Beverages
Legal InformationOmegawax is a registered trademark of Sigma-Aldrich Co. LLC
Other NotesComplementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
Suitabilityapplication for GC
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch