MilliporeSigma
HomeProtein Pull-Down TechniquesSera-Mag™ SpeedBeads Protocol
Sera-Mag™ SpeedBeads Protocol
Introduction
Important information before using Sera-Mag SpeedBeads Protein A/G Magnetic Particles
Procedure for manual antibody purification
Procedure for automated antibody purification
Procedure for manual immunoprecipitation (IP)
Procedure for automated immunoprecipitation
Troubleshooting
Materials
Protein enrichment
Sera-Mag SpeedBeads Protein A/G Magnetic Particles (Table 1) provide a fast and convenient method for both manual and automated magnetic isolation of proteins using affinity binding. The particles can be used for isolating antibodies from serum, cell culture supernatant or ascites, and for immunoprecipitation and co-immunoprecipitation of antigens from cell or tissue extracts. Bound antibodies or antigens are dissociated from the particles using an elution buffer.
The particles can be manually removed from the solution using a magnetic stand, or by automation using automated magnetic particle handling systems.
CompositionRecombinant protein A/G covalently coupled to particle surface
Magnetization
Superparamagnetic (no magnetic memory)
Mean diameter
1 μm (nomimal)
Concentration
10 mg/mL
Binding capacity55-85 μg IgG bound per mg of particle
Particle density~ 2.0 g/cm3
Table 1Characteristics of Sera-Mag SpeedBeads Protein A/G Magnetic Particles
Sera-Mag SpeedBeads Protein A/G Magnetic Particles contain a recombinant Protein A/G (Mr ~50 500; apparent molecular weight by SDS-PAGE Mr ~40,000 to 45,000) that combines the IgG binding domains of both Protein A and Protein G.
Protein A/G contains four Fc-binding domains from Protein A and two from Protein G, making it a more general and convenient tool for investigating and purifying immunoglobulins. Also, Protein A/G binding to immunoglobulins is not as pH-dependant as Protein A.
Sera-Mag SpeedBeads Protein A/G particles are uniform, colloidally stable, monodispersed, non-porous superparamagnetic spheres made by a proprietary core-shell method.
The core is a carboxylate-modified particle made by free radical emulsion polymerization of styrene and acid monomer. Two layers of magnetite (Fe3O4) are coated onto this core particle, resulting in faster magnetic response times. The surface is chemically modified with a proprietary method to minimize nonspecific binding of proteins. Finally, Protein A/G is covalently bound to the particle surface.
The particles are supplied at 1% solids (10 mg/mL) in 0.05% sodium azide and are available in 1 mL, 5 ml and 100 mLpackage sizes.
Important information before using Sera-Mag SpeedBeads Protein A/G Magnetic Particles
  • Do not centrifuge, dry or freeze the magnetic particles. Centrifuging, drying or freezing will cause the particles to aggregate and lose binding activity.
  • We recommend thoroughly mixing (vortex, roll or sonicate) magnetic protein A/G particles before use. Sonication is the preferred method to resuspend the particles thoroughly and efficiently.
  • Sonication with a probe type ultrasonicator is recommended to resuspend particles after long term storage, and washing steps.
  • To minimize protein degradation, include protease inhibitors in preparation of cell lysates.
  • A low-pH elution may be used for single-use applications. Optimal time for low-pH elution is 10 min. Exceeding 10 min may result in nonspecific binding and yield reduction.
  • When using rabbit antibodies (primary or secondary) in downstream Western blot applications, perform elution in SDS-PAGE sample buffer at room temperature. For all other antibody species, boiling the particles in SDS-PAGE sample buffer is acceptable for single-use applications. Boiling will cause particle aggregation and loss of binding activity.
  • Sera-Mag SpeedBeads Protein A/G Magnetic Particles are compatible with small-scale antibody purification and immunoprecipitation and analyses by Western blot and mass spectrometry.
Procedure for manual antibody purification
Additional materials required
  • 1.5 mL microcentrifuge tubes.
  • Sample: serum, concentrated cell culture supernatant or concentrated ascites.
  • Binding/Wash buffer: Tris-buffered saline (25 mM Tris, 0.15 M NaCl, pH 7.5) containing 0.05 % Tween®-20 detergent.
  • Elution buffer: 0.1 M glycine, pH 2-3.
  • Neutralization buffer: High-ionic strength alkaline buffer such as a 1 M phosphate or 1 M Tris; pH 7.5-9.
  • Magnetic stand (e.g., MagRack 6, Cytiva code number 28-9489-64).
Antibody purification from serum, cell culture supernatant, or ascites
Note: To ensure homogeneity, mix the particles thoroughly before use by repeated inversion, gentle vortexing or using a rotating platform.
  1. Place 50 μL (0.50 mg) of Sera-Mag SpeedBeads Protein A/G Magnetic Particles into a 1.5 mL microcentrifuge tube. Add 150 μL of binding/wash buffer to the particles and gently vortex to mix.
  2. Place the tube into a magnetic stand to collect the particles against the side of the tube. Remove and discard the supernatant.
  3. Add 1 mL of binding/wash buffer to the tube. Invert the tube several times or gently vortex to mix for 1 min. Collect particles with magnetic stand, then remove and discard the supernatant.
  4. Dilute 10 μL of sample with 490 μL binding/wash buffer.
    Note: Sample volume can be modified according to user preference. If the sample is < 500 μL dilute it to a final volume of 500 μL with binding/wash buffer.
  5. Add the diluted sample to the tube containing prewashed magnetic particles and gently vortex or invert to mix.
  6. Incubate the samples at room temperature with mixing for 1 h.
  7. Collect the particles with a magnetic stand, then remove and discard the supernatant.
  8. Add 500 μLof binding/wash buffer to the tube, mix well, collect the particles with a magnetic stand and discard the supernatant. Repeat this wash twice.
  9. Add 100 μL of elution buffer to the tube, mix well and incubate 10 min at room temperature with occasional mixing.
  10. Collect the particles with a magnetic stand and then remove and save the supernatant that contains the eluted antibody. To neutralize the low pH, add 10 μLof neutralization buffer for each 100 μL of eluate.
    Note: 50 μL is the minimum volume of particles recommended for antibody purification.
Procedure for automated antibody purification
Additional materials required
  • KingFisher™ Flex with 96 Deep Well head (Thermo Scientific product number 5400630) or KingFisher 96 (Thermo Scientific product number 5400500).
  • KingFisher Flex Microtiter Deepwell 96 plate, V-bottom (Thermo Scientific product number 95040450).
  • KingFisher Flex 96 tip comb for Deep Well Magnets (Thermo Scientific product number 97002534).
  • Binding/wash buffer: Tris-buffered saline containing 0.05% Tween®-20 detergent).
  • Elution buffer: 0.1 M glycine, pH 2-3.
  • Neutralization buffer: High-ionic strength alkaline buffer such as a 1 M phosphate or 1 M Tris; pH 7.5-9.
Preparation of instrument and plate set-up
Note: The following protocol is designed for general use with the KingFisher Flex or KingFisher 96 Instrument. The protocol can be modified according to customer needs using the Thermo Scientific BindIt™ software provided with the instrument.
  1. Download the “Antibody Purification” protocol from the Thermo Fisher Scientific website (www.thermoscientific.com/kingfisher) into the BindIt software on an external computer.
  2. Transfer the protocol to the KingFisher Flex or KingFisher 96 from an external computer. See BindIt Software User Manual for detailed instructions on importing protocols.
  3. Set up the plates according to Table 2.
Plate #Plate NameContentVolume
1ParticlesProtein A/G particles
Binding/Wash buffer
50 μL
150 μL
2Particle washBinding/Wash buffer1,000 μL
3BindSample
Binding/Wash buffer
10 μL
490 μL
4Wash 1Binding/Wash buffer500 μL
5Wash 2
Binding/Wash buffer500 μL
6Wash 3
Binding/Wash buffer500 μL
7ElutionElution buffer100 μL
8Tip plateKingFisher Flex 96 tip comb
for Deep Well magnets
-
Table 2Pipetting Instructions for the Antibody Protocol Using the Microtiter Deep Well 96 Plates
Notes
  • If using less than 96 wells, fill the same wells in each plate. For example, if using wells A1 through A12, use these same wells in all plates.
  • To ensure bead homogeneity, mix the vial thoroughly by repeated inversion, gentle vortexing or rotating platform before adding the particles to plate 1.
  • Combine the Tip Comb with a Deep Well 96 plate. See KingFisher Flex or KingFisher 96 user manual for detailed instructions.
  • Sample volume can be modified according to user preference. If the sample volume is < 500 μL dilute it to a final volume of 500 μL with binding/wash buffer.
Executing the antibody purification protocol on the KingFisher Flex
  1. Select the protocol using the arrows on the instrument key pad and press Start. See KingFisher Flex User Manual for detailed information.
  2. Slide open the door of the instrument’s protective cover.
  3. Load the plates into the KingFisher Flex according to the protocol request, placing each plate in the same orientation. Confirm each action by pressing Start.
  4. After the samples are processed, remove the plates as instructed by the instrument’s display. Press Start after removing each plate.
  5. Press Stop after all plates are removed.
  6. Upon completion, if desired, neutralize the low pH by adding 10 μL of neutralization buffer for each 100 μL of eluate.
Procedure for manual immunoprecipitation (IP)
Additional materials required
  • 1.5 mL microcentrifuge tubes.
  • Binding buffer: Tris-buffered saline (25 mM Tris, 0.15 M NaCl, pH 7.5) containing 0.05% Tween®-20 detergent.
  • Wash buffer: 25 mM Tris, 0.65 M NaCl, 0.05% Tween®-20 detergent, pH 7.5.
  • Low-pH elution buffer: 0.1 M glycine, pH 2-3.
  • Alternative elution buffer: SDS-PAGE reducing sample buffer.
  • Antibody for immunoprecipitation.
  • Antigen sample.
  • Cell lysis buffer (used to adjust IP reaction volume).
  • Neutralization buffer: High-ionic strength alkaline buffer such as a 1 M phosphate or 1 M Tris; pH 7.5-9.
Immunoprecipitation
Note: This protocol is a general guideline for immunoprecipitation and will require optimization for each application.
  1. Combine the antigen sample with 10 μg of antibody. Adjust the reaction volume to 500 μL with the cell lysis buffer. Incubate the reaction for 1 to 2 h at room temperature or overnight at 4ºC with mixing.
  2. Place 25 μL (0.25 mg) of Sera-Mag SpeedBeads Protein A/G Magnetic Particles into a 1.5 mL microcentrifuge tube.
  3. Add 175 μL of wash buffer to the particles and gently vortex to mix.
  4. Place the tube into a magnetic stand to collect the particles against the side of the tube. Remove and discard the supernatant.
  5. Add 1 mL of wash buffer to the tube. Invert the tube several times or gently vortex to mix for 1 min. Collect particles with magnetic stand. Remove and discard the supernatant.
  6. Add the antigen sample/antibody mixture to a 1.5 mL microcentrifuge tube containing prewashed particles and incubate at room temperature for 1 h with mixing.
  7. Collect the particles with a magnetic stand and then remove the flow-through and save for analysis.
  8. Add 500 μL of wash buffer to the tube and gently mix Collect the particles and then discard the supernatant. Repeat this wash twice.
  9. Add 500 μL of purified water to the tube and gently mix. Collect the particles on a magnetic stand and discard the supernatant.
  10. Low-pH Elution: Add 100 μL of low-pH elution buffer to the tube. Incubate the tube at room temperature with mixing for 10 min. Magnetically separate the particles and save the supernatant containing target antigen. To neutralize the low pH, add 10 μL of neutralization buffer for each 100 μL of eluate.
    Alternative elution: Add 100 μL of SDS-PAGE reducing sample buffer to the tube and heat the samples at 96 °C to 100 ºC in a heating block for 10 min. Magnetically separate the particles and save the supernatant containing target antigen.
    Note: If you will be performing a Western blot using rabbit antibodies (primary or secondary) do not heat the samples. Incubate at room temperature for 10 min with mixing.
Procedure for automated immunoprecipitation
Additional materials required
  • KingFisher Flex with 96 Deep Well head or KingFisher 96 (Thermo Scientific product no. 5400630 or 5400500)
  • KingFisher Flex Microtiter Deepwell 96 plate, V-bottom (Thermo Scientific product no. 95040450)
  • KingFisher Flex 96 tip comb for Deep Well magnets (Thermo Scientific product no. 97002534)
  • 1.5 mL microcentrifuge tubes
  • Binding buffer: Tris-buffered saline containing 0.05 % Tween®-20 detergent
  • Wash buffer: Tris-buffered saline containing 0.05 % Tween®-20 detergent and 0.5 M NaCl
  • Low-pH elution buffer: 0.1 M glycine, pH 2-3
  • Alternative elution buffer: SDS-PAGE reducing sample buffer.
  • Antigen sample
  • Cell lysis buffer (used to prepare the antigen sample)
  • Neutralization buffer: High-ionic strength alkaline buffer such as a 1 M phosphate or 1 M Tris; pH 7.5-9)
  • Magnetic stand 
Instrument preparation and plate set-up
Note: The following protocol is designed for general use with the KingFisher Flex or KingFisher 96 Instrument. The protocol can be modified according to your needs using the BindIt Software provided with the instrument.
  1. Combine antigen sample with 2-10 μg of immunoprecipitation antibody per sample. Incubate 1-2 h at room temperature or overnight at 4ºC with mixing.
  2. Enter the “Immunoprecipitation” protocol from Table 3 into the BindIt Software on an external computer.
  3. Transfer the protocol to the KingFisher Flex or KingFisher 96 from an external computer. See BindIt Software User Manual for detailed instructions on importing protocols.
  4. Set up plates according to Table 3.
Plate #Plate nameContentVolumeTime/Speed
1ParticlesProtein A/G particles
Binding buffer
50 μL
150 μL
5 s
2Particle washBinding buffer1,000 μL1 min/slow
3BindAntibody/
Antigen sample
500 μL1 min/slow
4Wash 1Binding/Wash buffer500 μL30 s/slow
5Wash 2
Binding/Wash buffer500 μL30 s/slow
6Wash 3
Ultrapure water500 μL30 s/slow
7ElutionElution buffer100 μL10 min/slow
8Tip plateKingFisher Flex 96 tip
comb for Deep Well
magnets
-10 min/fast
Table 3Pipetting instructions for the immunoprecipitation protocol using the Microtiter Deep Well 96 Plates
Notes
  • If using less than 96 wells, fill the same wells in each plate. For example, if using wells A1 through A12, use these same wells in all plates.
  • To ensure particle homogeneity, mix the vial thoroughly by repeated inversion, gentle vortexing or rotating platform before adding the particles to plate 1.
  • Combine the Tip Comb with a Deep Well 96 plate. See KingFisher Flex or KingFisher 96 user manual for detailed instructions.
  • The particles can be eluted into 100 μL of 0.1 M glycine, pH 2-3 or 100 μL SDS-PAGE reducing sample buffer. If using SDS-PAGE reducing sample buffer in a heated elution, install the KingFisher Flex or 96 Heating Block (see manual for proper installation) to heat samples at 96°C to 100 ºC for 10 min.
  • If you select SDS-PAGE reducing sample buffer for elution and will be performing a Western blot using rabbit antibodies (primary or secondary), do not heat the samples. Incubate at room temperature for 10 min.
  • If low-pH elution buffer is selected for elution, neutralize the pH using 10 μL neutralization buffer for each 100 μL of eluate upon run completion.
  • To limit evaporation, select “Mix” and “Slow” speed under the subheading “Heating Action”.
Executing automated immunoprecipitation protocol
  1. Select the protocol using the arrows on the instrument key pad and press Start. See the KingFisher Flex User or Kingfisher 96 User Manual for detailed information.
  2. Slide open the door of the instrument’s protective cover.
  3. Load the plates into the instrument according to the protocol request, placing each plate in the same orientation. Confirm each action by pressing Start.
  4. After the samples are processed, remove the plates as instructed by the instrument’s display. Press Start after removing each plate. Press Stop after all the plates are removed.
Troubleshooting
ProblemPossible CauseSolution
Low amount of protein was recoveredThe protein degraded
Insufficient magnetic particles used Insufficient target protein present in sample
Add protease inhibitors Increase the amount of magnetic particles used for capture Increase amount of antigen sample
Protein does not eluteElution conditions are too mildIncrease incubation time with elution buffer or use more stringent elution buffer
Bands at Mr 50 000 appear on Western blotElution conditions are too stringentPerform elution at room temperature
Multiple, nonspecific
bands appear in eluted sample
Nonspecific protein binding to the magnetic particlesAdd 50-200 mM NaCl to the binding/wash and/or elution buffers
Recovered protein was inactiveElution conditions are too stringentUse a milder elution buffer
Magnetic particles
aggregate
Magnetic particles were frozen or centrifuged Buffer is incompatible with magnetic particlesHandle the particles as directed in the instructions
Table 4.General troubleshooting tips and suggestions for Sera-Mag Speedbeads Protein A/G Magnetic Particles
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch